Gene Transcripts

Gene Symbol
RIPK1
Entrez Gene ID
8737
Gene Name
receptor interacting serine/threonine kinase 1
Gene Aliases
RIP, RIP-1, RIP1
Chromosome Location
Chr. 6: 3063888 - 3115187 on Build GRCh38
UniGene ID
Hs.519842
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317061.1 NP_001303990.1 7 1058
NM_003804.4 NP_003795.2 5 780
XM_005249457.3 XP_005249514.2 5 703
XM_006715237.3 XP_006715300.2 5 703
XM_017011403.1 XP_016866892.1 2 812
XM_017011405.1 XP_016866894.1 5 701
GenBank mRNA AB208926.1 - 7 967
AK304701.1 - 3 369
AK314176.1 - 5 780
BC126254.1 - 4 574 8991970
BC126256.1 - 4 574 8991972
U50062.1 - 4 548

View Full Product Details