Gene Transcripts

Gene Symbol
BTAF1
Entrez Gene ID
9044
Gene Name
B-TFIID TATA-box binding protein associated factor 1
Gene Aliases
MOT1, TAF(II)170, TAF172, TAFII170
Chromosome Location
Chr. 10: 91923769 - 92030998 on Build GRCh38
UniGene ID
Hs.500526
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003972.2 NP_003963.1 17, 18 2150
XM_011540326.1 XP_011538628.1 17, 18 2249
XM_011540327.2 XP_011538629.1 14, 15 1915
XM_011540328.2 XP_011538630.1 14, 15 1743
XM_011540329.2 XP_011538631.1 17, 18 2360
XM_017016877.1 XP_016872366.1 16, 17 2212
XM_017016878.1 XP_016872367.1 16, 17 2042
XM_017016879.1 XP_016872368.1 16, 17 2166
XM_017016880.1 XP_016872369.1 16, 17 2170
XM_017016881.1 XP_016872370.1 15, 16 2019
GenBank mRNA AB208843.1 - 4, 5 439
AF038362.1 - 17, 18 2169
AJ001017.2 - 17, 18 2064
BC030273.1 - 11, 12 1397
BC047009.2 - 5, 6 642
BC112201.1 - 17, 18 2134 8327669

View Full Product Details