Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005113.3 | NP_005104.3 | 2 | 460 | |
XM_011537420.2 | XP_011535722.1 | 2 | 460 | ||
GenBank mRNA | AF085199.1 | - | 2 | 335 | |
AJ132949.1 | - | 1 | 204 | ||
AK293077.1 | - | 2 | 363 | ||
AK303789.1 | - | 2 | 366 | ||
AL702150.1 | - | 2 | 388 | ||
AY644768.1 | - | 1 | 240 |