Gene Transcripts

Gene Symbol
PAGE4
Entrez Gene ID
9506
Gene Name
PAGE family member 4
Gene Aliases
CT16.7, GAGE-9, GAGEC1, JM-27, JM27, PAGE-1, PAGE-4
Chromosome Location
Chr. X: 49829260 - 49834264 on Build GRCh38
UniGene ID
Hs.441038
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318877.1 NP_001305806.1 3, 4 244
NM_007003.4 NP_008934.1 3, 4 256
GenBank mRNA AF275258.1 - 3, 4 242
AJ005894.1 - 3, 4 240
AK312039.1 - 3, 4 253
AL548759.3 - 2, 3 207
BC010897.2 - 2, 3 206 4043535
BG435968.1 - 3, 4 257
BG622311.1 - 258
CR456823.1 - 2, 3 177
CR542183.1 - 2, 3 177
DA841276.1 - 3, 4 238

View Full Product Details