Gene Transcripts

Gene Symbol
THBS2
Entrez Gene ID
7058
Gene Name
thrombospondin 2
Gene Aliases
TSP2
Chromosome Location
Chr. 6: 169215780 - 169254114 on Build GRCh38
UniGene ID
Hs.371147
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003247.3 NP_003238.2 4 402
GenBank mRNA AK292429.1 - 4 359
BC146676.1 - 3 209 40148868
BC150175.1 - 3 213 40147958
BX390187.2 - 2 74
DB291377.1 - 4 403
L12350.1 - 4 321

View Full Product Details