Gene Transcripts

Gene Symbol
CDK13
Entrez Gene ID
8621
Gene Name
cyclin dependent kinase 13
Gene Aliases
CDC2L, CDC2L5, CHED, hCDK13
Chromosome Location
Chr. 7: 39949501 - 40097134 on Build GRCh38
UniGene ID
Hs.233552
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003718.4 NP_003709.3 10 3152
NM_031267.3 NP_112557.2 10 3152
XM_017012750.1 XP_016868239.1 11 4101
XM_017012751.1 XP_016868240.1 11 4101
GenBank mRNA AB058694.1 - 4 2568
AJ297709.1 - 10 3002
AJ297710.1 - 10 3002
AK223053.1 - 4 305
BF707421.1 - 3 199
M80629.1 - 10 1799

View Full Product Details