Gene Transcripts
- Gene Symbol
- TIE1
- Entrez Gene ID
- 7075
- Gene Name
- tyrosine kinase with immunoglobulin like and EGF like domains 1
- Gene Aliases
- JTK14, TIE
- Chromosome Location
- Chr. 1: 43300895 - 43323110 on Build GRCh38
- UniGene ID
- Hs.78824
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001253357.1 |
NP_001240286.1
|
16, 17 |
2833 |
|
NM_005424.4 |
NP_005415.1
|
16, 17 |
2916 |
|
XM_005271163.1 |
XP_005271220.1
|
15, 16 |
2730 |
|
XM_006710869.1 |
XP_006710932.1
|
16, 17 |
3050 |
|
GenBank mRNA |
AF217976.1 |
- |
6, 7 |
1000 |
|
AK225560.1 |
- |
16, 17 |
2575 |
|
AK296735.1 |
- |
11, 12 |
1892 |
|
AK300399.1 |
- |
16, 17 |
2723 |
|
AK307639.1 |
- |
9, 10 |
1584 |
|
BC038239.1 |
- |
16, 17 |
2818 |
5767075 |
X60957.1 |
- |
16, 17 |
2775 |
|
View Full Product Details