Gene Transcripts

Gene Symbol
RGS11
Entrez Gene ID
8786
Gene Name
regulator of G-protein signaling 11
Gene Aliases
RS11
Chromosome Location
Chr. 16: 268301 - 275943 on Build GRCh38
UniGene ID
Hs.65756
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286485.1 NP_001273414.1 15 1227
NM_001286486.1 NP_001273415.1 14 1095
NM_003834.2 NP_003825.1 16 1271
NM_183337.2 NP_899180.1 16 1260
XM_011522719.2 XP_011521021.1 17 1354
XM_011522720.2 XP_011521022.1 17 1294
GenBank mRNA AB016929.1 - 15 1195
AB209582.1 - 8 3168
AF035153.1 - 16 1231
AF035154.1 - 16 1242
AK091701.1 - 14 1092
AK293564.1 - 16 1141
BC141803.1 - 16 1235 40147243

View Full Product Details