Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001122957.2 | NP_001116429.1 | 4 | 656 | |
NM_001271926.1 | NP_001258855.1 | 4 | 656 | ||
NM_005881.3 | NP_005872.2 | 4 | 656 | ||
XM_017022859.1 | XP_016878348.1 | 4 | 569 | ||
GenBank mRNA | AF026548.1 | - | 4 | 566 | |
AK130145.1 | - | 4 | 536 | ||
AK294691.1 | - | 3 | 365 | ||
BC007363.2 | - | 4 | 539 | 3630050 | |
BC009872.2 | - | 4 | 519 | 3944953 | |
BX427330.2 | - | 4 | 653 | ||
CR542093.1 | - | 3 | 293 |