Gene Transcripts
- Gene Symbol
- SNF8
- Entrez Gene ID
- 11267
- Gene Name
- SNF8, ESCRT-II complex subunit
- Gene Aliases
- Dot3, EAP30, VPS22
- Chromosome Location
- Chr. 17: 48930096 - 48944842 on Build GRCh38
- UniGene ID
- Hs.127249
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001317192.1 |
NP_001304121.1
|
4, 5 |
465 |
|
NM_001317193.1 |
NP_001304122.1
|
3, 4 |
414 |
|
NM_007241.3 |
NP_009172.2
|
4, 5 |
465 |
|
NR_133679.1 |
|
4, 5 |
465 |
|
GenBank mRNA |
AF156102.1 |
- |
4, 5 |
414 |
|
BC008976.1 |
- |
4, 5 |
415 |
3452153 |
BC038830.1 |
- |
4, 5 |
439 |
3900806 |
BE818533.1 |
- |
3, 4 |
268 |
|
BG248541.1 |
- |
4, 5 |
454 |
|
BP342490.1 |
- |
4, 5 |
465 |
|
View Full Product Details