Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016073.3 | NP_057157.1 | 5, 6 | 1208 | |
GenBank mRNA | AB029156.1 | - | 5, 6 | 724 | |
AF110642.1 | - | 5, 6 | 728 | ||
AF151900.1 | - | 5, 6 | 728 | ||
AK001280.1 | - | 5, 6 | 759 | ||
AL109689.1 | - | 1, 2 | 125 | ||
BC015483.1 | - | 5, 6 | 702 | 3913558 | |
BC040554.1 | - | 5, 6 | 1211 |