Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016125.3 | NP_057209.3 | 8 | 1192 | |
GenBank mRNA | AK098649.1 | - | 6 | 572 | |
AK296197.1 | - | 6 | 995 | ||
BC006971.1 | - | 7 | 1069 | ||
BC088365.1 | - | 8 | 1155 | 30390062 |