Gene Transcripts

Gene Symbol
ANKRD40
Entrez Gene ID
91369
Gene Name
ankyrin repeat domain 40
Gene Aliases
Chromosome Location
Chr. 17: 50693190 - 50707909 on Build GRCh38
UniGene ID
Hs.463426
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_052855.3 NP_443087.1 3 959
XM_017025296.1 XP_016880785.1 3 706
GenBank mRNA AK314010.1 - 3 706
BC012978.2 - 3 951 3834331
CR627413.1 - 3 731
EU164539.1 - 3 951

View Full Product Details