Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_138573.3 | NP_612640.1 | 4 | 380 | |
XM_017021937.1 | XP_016877426.1 | 8 | 891 | ||
XM_017021938.1 | XP_016877427.1 | 7 | 3098 | ||
XM_017021939.1 | XP_016877428.1 | 7 | 660 | ||
XM_017021940.1 | XP_016877429.1 | 6 | 3436 | ||
XM_017021941.1 | XP_016877430.1 | 4 | 399 | ||
XM_017021942.1 | XP_016877431.1 | 4 | 417 | ||
XM_017021943.1 | XP_016877432.1 | 4 | 490 | ||
XM_017021944.1 | XP_016877433.1 | 7 | 3483 | ||
XM_017021945.1 | XP_016877434.1 | 7 | 3475 | ||
XM_017021946.1 | XP_016877435.1 | 7 | 3475 | ||
XM_017021947.1 | XP_016877436.1 | 7 | 3475 | ||
XM_017021948.1 | XP_016877437.1 | 5 | 3208 | ||
GenBank mRNA | AK307362.1 | - | 4 | 358 | |
AK311204.1 | - | 9 | 712 | ||
AK311507.1 | - | 5 | 482 | ||
AM392364.1 | - | 2 | 198 | ||
BC017568.1 | - | 4 | 384 | 4071605 | |
BX538100.1 | - | 5 | 410 | ||
CR749519.1 | - | 6 | 554 |