Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001308025.1 | NP_001294954.1 | 20 | 3888 | |
NM_152259.3 | NP_689472.3 | 20 | 3891 | ||
GenBank mRNA | AK074727.1 | - | 1 | 383 | |
AK123612.1 | - | 13 | 1991 | ||
AK315750.1 | - | 1 | 203 | ||
BX538335.1 | - | 7 | 1192 | ||
GQ227787.1 | - | 20 | 3888 |