Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_174892.3 | NP_777552.3 | 3 | 527 | |
XM_005257027.3 | XP_005257084.1 | 3 | 527 | ||
GenBank mRNA | AF427618.1 | - | 3 | 427 | |
AK290925.1 | - | 3 | 527 | ||
AK298665.1 | - | 3 | 528 | ||
AY359025.1 | - | 3 | 456 | ||
AY646929.1 | - | 3 | 474 | ||
BC028091.1 | - | 3 | 519 | 5219239 | |
DA673717.1 | - | 3 | 529 |