Gene Transcripts

Gene Symbol
KRT40
Entrez Gene ID
125115
Gene Name
keratin 40
Gene Aliases
CK-40, K40, KA36
Chromosome Location
Chr. 17: 40977716 - 40987135 on Build GRCh38
UniGene ID
Hs.534929
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182497.3 NP_872303.2 9 1658
XM_017024189.1 XP_016879678.1 7 3425
GenBank mRNA AJ786658.1 - 7 1498
AK093919.1 - 9 1832
BC150174.1 - 9 1658 40147687
BC150191.1 - 9 1658 40148622

View Full Product Details