Gene Transcripts

Gene Symbol
ROS1
Entrez Gene ID
6098
Gene Name
ROS proto-oncogene 1, receptor tyrosine kinase
Gene Aliases
MCF3, ROS, c-ros-1
Chromosome Location
Chr. 6: 117287918 - 117426065 on Build GRCh38
UniGene ID
Hs.1041
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002944.2 NP_002935.2 7 877
XM_006715548.3 XP_006715611.1 8 1011
XM_011536049.2 XP_011534351.1 8 1012
XM_011536050.2 XP_011534352.1 8 1012
XM_011536051.2 XP_011534353.1 7 985
XM_011536052.2 XP_011534354.1 8 1012
XM_011536053.2 XP_011534355.1 8 1012
XM_011536054.2 XP_011534356.1 8 1013
XM_011536055.2 XP_011534357.1 8 1012
XM_011536056.2 XP_011534358.1 8 1011
XM_011536057.2 XP_011534359.1 8 1010
XM_011536058.2 XP_011534360.1 8 1010
XM_017011172.1 XP_016866661.1 7 985
XM_017011173.1 XP_016866662.1 7 1087
GenBank mRNA BC144556.1 - 7 725
M34353.1 - 7 884

View Full Product Details