Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005408.2 | NP_005399.1 | 3 | 605 | |
GenBank mRNA | AJ001634.1 | - | 3 | 572 | |
BC008621.1 | - | 3 | 568 | 4184250 | |
U46767.1 | - | 3 | 563 | ||
U59808.1 | - | 3 | 605 | ||
Z77650.1 | - | 3 | 481 | ||
Z77651.1 | - | 2 | 423 |