Gene Transcripts
- Gene Symbol
- FLT3
- Entrez Gene ID
- 2322
- Gene Name
- fms related tyrosine kinase 3
- Gene Aliases
- CD135, FLK-2, FLK2, STK1
- Chromosome Location
- Chr. 13: 28003274 - 28100592 on Build GRCh38
- UniGene ID
- Hs.507590
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_004119.2 |
NP_004110.2
|
10, 11 |
1385 |
|
NR_130706.1 |
|
10, 11 |
1385 |
|
XM_011535015.2 |
XP_011533317.1
|
10, 11 |
1384 |
|
XM_011535017.2 |
XP_011533319.1
|
10, 11 |
1287 |
|
XM_011535018.2 |
XP_011533320.1
|
7, 8 |
895 |
|
XM_017020486.1 |
XP_016875975.1
|
9, 10 |
1164 |
|
XM_017020487.1 |
XP_016875976.1
|
11, 12 |
1458 |
|
XM_017020488.1 |
XP_016875977.1
|
9, 10 |
1151 |
|
XM_017020489.1 |
XP_016875978.1
|
9, 10 |
1246 |
|
GenBank mRNA |
BC036028.1 |
- |
10, 11 |
1385 |
|
BC126350.1 |
- |
10, 11 |
1369 |
8992066 |
BC144039.1 |
- |
10, 11 |
1370 |
9052559 |
BC144040.1 |
- |
10, 11 |
1370 |
9052560 |
U02687.1 |
- |
10, 11 |
1360 |
|
Z26652.1 |
- |
10, 11 |
1303 |
|
View Full Product Details