Gene Transcripts

Gene Symbol
CD101
Entrez Gene ID
9398
Gene Name
CD101 molecule
Gene Aliases
EWI-101, IGSF2, V7
Chromosome Location
Chr. 1: 116990423 - 117037481 on Build GRCh38
UniGene ID
Hs.654598
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256106.2 NP_001243035.1 7 2341
NM_001256109.2 NP_001243038.1 7 2341
NM_001256111.2 NP_001243040.1 7 2155
NM_004258.5 NP_004249.2 7 2341
XM_017002846.1 XP_016858335.1 8 2294
XM_017002847.1 XP_016858336.1 8 2294
GenBank mRNA AK298237.1 - 7 2130
BC130327.1 - 8 2302 40146358
BC136484.1 - 8 2302 9020473
BC171768.1 - 8 2302 9054422
Z33642.1 - 8 2294

View Full Product Details