Gene Transcripts
- Gene Symbol
- SEPT7
- Entrez Gene ID
- 989
- Gene Name
- septin 7
- Gene Aliases
- CDC10, CDC3, NBLA02942, SEPT7A
- Chromosome Location
- Chr. 7: 35800986 - 35915763 on Build GRCh38
- UniGene ID
- Hs.191346
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001011553.3 |
NP_001011553.2
|
5, 6 |
734 |
|
NM_001242956.1 |
NP_001229885.1
|
4, 5 |
629 |
|
NM_001788.5 |
NP_001779.3
|
6, 7 |
737 |
|
XM_006715806.3 |
XP_006715869.1
|
6, 7 |
685 |
|
XM_011515656.2 |
XP_011513958.1
|
6, 7 |
685 |
|
XM_011515661.2 |
XP_011513963.1
|
6, 7 |
685 |
|
XM_017012855.1 |
XP_016868344.1
|
6, 7 |
669 |
|
XM_017012856.1 |
XP_016868345.1
|
6, 7 |
677 |
|
XM_017012857.1 |
XP_016868346.1
|
6, 7 |
674 |
|
XM_017012858.1 |
XP_016868347.1
|
7, 8 |
599 |
|
XM_017012859.1 |
XP_016868348.1
|
6, 7 |
596 |
|
XM_017012860.1 |
XP_016868349.1
|
7, 8 |
593 |
|
XM_017012861.1 |
XP_016868350.1
|
7, 8 |
590 |
|
XM_017012862.1 |
XP_016868351.1
|
5, 6 |
500 |
|
XM_017012863.1 |
XP_016868352.1
|
6, 7 |
1043 |
|
GenBank mRNA |
AB073389.1 |
- |
6, 7 |
504 |
|
AB209677.1 |
- |
6, 7 |
472 |
|
AK223225.1 |
- |
6, 7 |
504 |
|
AK223241.1 |
- |
6, 7 |
504 |
|
AK290545.1 |
- |
5, 6 |
575 |
|
AK297883.1 |
- |
4, 5 |
396 |
|
AK298453.1 |
- |
6, 7 |
571 |
|
AK300601.1 |
- |
7, 8 |
450 |
|
BC025987.2 |
- |
5, 6 |
501 |
4823810 |
BC067264.1 |
- |
5, 6 |
480 |
6503495 |
BC093640.1 |
- |
6, 7 |
503 |
7939485 |
BC093642.1 |
- |
6, 7 |
503 |
7939487 |
BC107718.1 |
- |
5, 6 |
682 |
|
BQ229116.1 |
- |
5, 6 |
665 |
|
BQ232357.1 |
- |
6, 7 |
479 |
|
BX571755.1 |
- |
5, 6 |
502 |
|
DQ232879.1 |
- |
5, 6 |
485 |
|
KF305319.1 |
- |
7, 8 |
569 |
|
S72008.1 |
- |
6, 7 |
504 |
|
- Gene Symbol
- LOC441601
- Entrez Gene ID
- 441601
- Gene Name
- septin 7 pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 11: 50279828 - 50298462 on Build GRCh38
- UniGene ID
- Hs.144683
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003034.2 |
|
3, 4 |
526 |
|
GenBank mRNA |
BC067889.1 |
- |
3, 4 |
516 |
|
View Full Product Details