Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001888.4 | NP_001879.1 | 6 | 808 | |
GenBank mRNA | AK290852.1 | - | 4 | 527 | |
AK310424.1 | - | 4 | 481 | ||
BC018061.1 | - | 4 | 526 | 4817634 | |
BI860684.1 | - | 5 | 481 | ||
BX648477.1 | - | 6 | 724 | ||
L02950.1 | - | 4 | 487 | ||
U85772.1 | - | 4 | 482 |