Gene Transcripts

Gene Symbol
EPHB6
Entrez Gene ID
2051
Gene Name
EPH receptor B6
Gene Aliases
HEP
Chromosome Location
Chr. 7: 142854640 - 142871093 on Build GRCh38
UniGene ID
Hs.380089
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001280794.2 NP_001267723.2 6 958
NM_001280795.2 NP_001267724.2 3 330
NM_004445.5 NP_004436.4 4 399
NR_104001.2 5 516
XM_006715881.2 XP_006715944.1 7 1394
XM_011515879.1 XP_011514181.1 8 1433
XM_011515880.1 XP_011514182.1 7 1411
XM_011515881.2 XP_011514183.1 7 1395
XM_011515882.2 XP_011514184.1 7 1397
GenBank mRNA BC056159.1 - 6 830
BC110606.1 - 3 327 40028676
BC110607.1 - 4 396 40028687
BX410199.2 - 3 321
CN283348.1 - 1 21
D83492.1 - 7 973
EU054308.1 - 3 175

View Full Product Details