Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004629.1 | NP_004620.1 | 8 | 1541 | |
GenBank mRNA | AJ007669.1 | - | 8 | 1541 | |
AK293427.1 | - | 9 | 1231 | ||
AK311348.1 | - | 7 | 1015 | ||
AK312987.1 | - | 8 | 1466 | ||
BC000032.2 | - | 8 | 1187 | 3507258 | |
BC011623.2 | - | 8 | 1314 | 4053144 | |
BT009813.1 | - | 8 | 1049 | ||
KU159422.1 | - | 8 | 1572 | ||
KU178053.1 | - | 8 | 1049 | ||
KU178054.1 | - | 8 | 1049 | ||
KU178055.1 | - | 8 | 1049 | ||
KU178056.1 | - | 5 | 623 | ||
U70310.1 | - | 8 | 1183 |