Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004427.3 | NP_004418.2 | 5 | 776 | |
NM_198040.2 | NP_932157.1 | 12 | 2083 | ||
XM_005270570.1 | XP_005270627.1 | 5 | 712 | ||
XM_017000513.1 | XP_016856002.1 | 13 | 2582 | ||
XM_017000514.1 | XP_016856003.1 | 13 | 2194 | ||
XM_017000515.1 | XP_016856004.1 | 13 | 2287 | ||
XM_017000516.1 | XP_016856005.1 | 13 | 2516 | ||
XM_017000517.1 | XP_016856006.1 | 13 | 2128 | ||
XM_017000518.1 | XP_016856007.1 | 13 | 2221 | ||
XM_017000519.1 | XP_016856008.1 | 13 | 2591 | ||
XM_017000520.1 | XP_016856009.1 | 12 | 2041 | ||
XM_017000521.1 | XP_016856010.1 | 12 | 2460 | ||
GenBank mRNA | AJ419231.1 | - | 12 | 2083 | |
AK056429.1 | - | 6 | 1013 | ||
AK128821.1 | - | 5 | 594 | ||
AK292905.1 | - | 13 | 2521 | ||
AK294426.1 | - | 7 | 1361 | ||
BC018602.1 | - | 5 | 782 | ||
BC028396.1 | - | 5 | 747 | ||
BC029269.1 | - | 5 | 618 | ||
BC068573.1 | - | 5 | 670 | ||
BC092492.1 | - | 5 | 670 | ||
BC110863.1 | - | 5 | 669 | ||
BC130630.1 | - | 11 | 1999 | 40146661 | |
BC144121.1 | - | 11 | 1999 | 9052641 | |
U89278.1 | - | 5 | 768 |