Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004427.3 | NP_004418.2 | 2 | 478 | |
NM_198040.2 | NP_932157.1 | 9 | 1785 | ||
XM_005270570.1 | XP_005270627.1 | 2 | 414 | ||
XM_017000513.1 | XP_016856002.1 | 10 | 2284 | ||
XM_017000514.1 | XP_016856003.1 | 10 | 1896 | ||
XM_017000515.1 | XP_016856004.1 | 10 | 1989 | ||
XM_017000516.1 | XP_016856005.1 | 10 | 2218 | ||
XM_017000517.1 | XP_016856006.1 | 10 | 1830 | ||
XM_017000518.1 | XP_016856007.1 | 10 | 1923 | ||
XM_017000519.1 | XP_016856008.1 | 10 | 2293 | ||
XM_017000520.1 | XP_016856009.1 | 9 | 1743 | ||
XM_017000521.1 | XP_016856010.1 | 9 | 2162 | ||
XM_017000522.1 | XP_016856011.1 | 10 | 2323 | ||
GenBank mRNA | AJ419231.1 | - | 9 | 1785 | |
AK056429.1 | - | 3 | 715 | ||
AK292905.1 | - | 10 | 2223 | ||
AK294426.1 | - | 4 | 1063 | ||
BC018602.1 | - | 2 | 484 | ||
BC028396.1 | - | 2 | 449 | ||
BC029269.1 | - | 2 | 320 | ||
BC068573.1 | - | 2 | 372 | ||
BC092492.1 | - | 2 | 372 | ||
BC110863.1 | - | 2 | 371 | ||
BC130630.1 | - | 8 | 1701 | 40146661 | |
BC144121.1 | - | 8 | 1701 | 9052641 | |
CD109667.1 | - | 2 | 495 | ||
U89278.1 | - | 2 | 473 |