Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001715.2 | NP_001706.2 | 4 | 798 | |
XM_011543824.1 | XP_011542126.1 | 4 | 819 | ||
XM_011543825.2 | XP_011542127.1 | 4 | 564 | ||
XM_011543827.1 | XP_011542129.1 | 3 | 695 | ||
XM_011543828.2 | XP_011542130.1 | 4 | 819 | ||
XM_011543829.2 | XP_011542131.1 | 4 | 819 | ||
XM_017013730.1 | XP_016869219.1 | 3 | 695 | ||
GenBank mRNA | AK313751.1 | - | 4 | 387 | |
BC007371.2 | - | 4 | 361 | 3634568 | |
BC018703.1 | - | 4 | 662 | ||
BC032413.1 | - | 4 | 407 | 5216030 | |
S76617.1 | - | 4 | 805 | ||
Z33998.1 | - | 4 | 439 |