Gene Transcripts

Gene Symbol
BMP8B
Entrez Gene ID
656
Gene Name
bone morphogenetic protein 8b
Gene Aliases
BMP8, OP2
Chromosome Location
Chr. 1: 39757182 - 39788861 on Build GRCh38
UniGene ID
Hs.734367
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001720.3 NP_001711.2 7 1718
XM_011542024.2 XP_011540326.2 8 1511
GenBank mRNA AY303955.1 - 7 1718
BC023526.2 - 7 1317

View Full Product Details