Gene Transcripts

Gene Symbol
EML1
Entrez Gene ID
2009
Gene Name
echinoderm microtubule associated protein like 1
Gene Aliases
ELP79, EMAP, EMAPL, HuEMAP
Chromosome Location
Chr. 14: 99737699 - 99942060 on Build GRCh38
UniGene ID
Hs.12451
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008707.1 NP_001008707.1 9, 10 1022
NM_004434.2 NP_004425.2 8, 9 965
XM_005267397.1 XP_005267454.1 9, 10 1092
XM_005267398.3 XP_005267455.1 9, 10 1025
XM_005267399.4 XP_005267456.2 9, 10 1064
XM_005267400.4 XP_005267457.1 8, 9 991
XM_011536540.2 XP_011534842.1 8, 9 1038
XM_011536541.1 XP_011534843.1 9, 10 1105
XM_011536542.2 XP_011534844.1 9, 10 1064
XM_017021074.1 XP_016876563.1 8, 9 1035
GenBank mRNA AK126980.1 - 10, 11 978
AK299812.1 - 9, 10 1301
AK307925.1 - 8, 9 958
BC032541.1 - 8, 9 941 5533599
BC033043.1 - 8, 9 965 4821202
BX247979.1 - 9, 10 1034
U97018.1 - 8, 9 964

View Full Product Details