Gene Transcripts

Gene Symbol
C1S
Entrez Gene ID
716
Gene Name
complement component 1, s subcomponent
Gene Aliases
Chromosome Location
Chr. 12: 7060676 - 7071032 on Build GRCh38
UniGene ID
Hs.458355
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001734.3 NP_001725.1 12 2033
NM_201442.2 NP_958850.1 12 2120
XM_005253760.1 XP_005253817.1 12 2012
GenBank mRNA AK025309.1 - 9 1508
AK290295.1 - 12 2067
AM886411.1 - 11 1755
BC056903.1 - 12 2060 6199186
J04080.1 - 12 1959
M18767.1 - 12 1979

View Full Product Details