Gene Transcripts

Gene Symbol
PTTG1IP
Entrez Gene ID
754
Gene Name
pituitary tumor-transforming 1 interacting protein
Gene Aliases
C21orf1, C21orf3, PBF
Chromosome Location
Chr. 21: 44849585 - 44873903 on Build GRCh38
UniGene ID
Hs.474010
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286822.1 NP_001273751.1 3 1790
NM_004339.3 NP_004330.1 6 2118
NR_104597.1 5 2009
GenBank mRNA AF149785.1 - 6 2062
AK094882.1 - 7 1823
AK095586.1 - 1 2419
BC000415.2 - 6 1876 2961420
BC012858.1 - 6 1901 3857505
BC019295.1 - 6 1865 2823814
BC020983.1 - 6 1882 3851447
BC031097.1 - 7 1845 5288388
BC034250.1 - 6 1890 3862830
L48984.1 - 3 1516
Z50022.1 - 6 1921

View Full Product Details