Gene Transcripts

Gene Symbol
RFTN1
Entrez Gene ID
23180
Gene Name
raftlin, lipid raft linker 1
Gene Aliases
MIG2, PIB10, PIG9, RAFTLIN
Chromosome Location
Chr. 3: 16315845 - 16513715 on Build GRCh38
UniGene ID
Hs.98910
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015150.1 NP_055965.1 5, 6 1103
XM_005264985.1 XP_005265042.1 5, 6 1060
XM_005264986.2 XP_005265043.1 5, 6 957
XM_006713069.1 XP_006713132.1 5, 6 966
XM_011533529.1 XP_011531831.1 7, 8 1500
XM_011533530.1 XP_011531832.1 5, 6 1128
XM_011533531.1 XP_011531833.1 7, 8 1500
XM_017006005.1 XP_016861494.1 7, 8 1500
XM_017006006.1 XP_016861495.1 7, 8 1500
XM_017006007.1 XP_016861496.1 5, 6 1087
GenBank mRNA AK074446.1 - 5, 6 909
AK301024.1 - 4, 5 796
AY237654.1 - 5, 6 1094
BC006400.2 - 1, 2 31
BC014032.2 - 1, 2 31
BC032349.1 - 4, 5 776
BC051336.1 - 5, 6 880
BC069209.1 - 5, 6 880
BC098270.1 - 5, 6 927 40001191
BC100667.1 - 5, 6 927 40001188
BC100669.1 - 5, 6 927 40001190
BC121795.1 - 5, 6 927 40011545
D42043.1 - 5, 6 1031

View Full Product Details