Gene Transcripts

Gene Symbol
BRD9
Entrez Gene ID
65980
Gene Name
bromodomain containing 9
Gene Aliases
LAVS3040, PRO9856
Chromosome Location
Chr. 5: 863735 - 892824 on Build GRCh38
UniGene ID
Hs.449278
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009877.2 NP_001009877.2 4 420
NM_001317951.1 NP_001304880.1 4 707
NM_023924.4 NP_076413.3 4 588
NR_134296.1 4 924
XM_006714494.3 XP_006714557.1 4 567
XM_017009780.1 XP_016865269.1 4 567
GenBank mRNA AK023503.1 - 4 685
AK024392.1 - 1 423
AK297573.1 - 4 407
AK299157.1 - 4 588
AK301987.1 - 4 539
AY211916.1 - 4 555
BC007217.1 - 3 431
BC041590.1 - 2 274 5428011
BX648703.1 - 4 926

View Full Product Details