Gene Transcripts

Gene Symbol
C10orf76
Entrez Gene ID
79591
Gene Name
chromosome 10 open reading frame 76
Gene Aliases
Chromosome Location
Chr. 10: 101845599 - 102056237 on Build GRCh38
UniGene ID
Hs.16004
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024541.2 NP_078817.2 26 2479
XM_006717969.1 XP_006718032.1 24 2267
XM_011540151.2 XP_011538453.1 26 2524
XM_011540152.2 XP_011538454.1 25 2411
XM_011540153.2 XP_011538455.1 25 2375
XM_011540154.2 XP_011538456.1 24 2309
XM_011540156.2 XP_011538458.1 19 2443
XM_011540157.2 XP_011538459.1 25 2428
GenBank mRNA AK023176.1 - 9 1108
AK097972.1 - 3 842

View Full Product Details