Gene Transcripts

Gene Symbol
SUSD3
Entrez Gene ID
203328
Gene Name
sushi domain containing 3
Gene Aliases
Chromosome Location
Chr. 9: 93058688 - 93085138 on Build GRCh38
UniGene ID
Hs.88417
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001287005.1 NP_001273934.1 5 675
NM_001287008.1 NP_001273937.1 3 372
NM_145006.3 NP_659443.1 4 561
XM_011518358.1 XP_011516660.1 7 1755
XM_011518359.1 XP_011516661.1 6 1705
XM_011518361.2 XP_011516663.1 7 817
XM_011518362.1 XP_011516664.1 5 1591
XM_017014448.1 XP_016869937.1 5 862
XM_017014449.1 XP_016869938.1 4 743
XM_017014450.1 XP_016869939.1 6 702
XM_017014451.1 XP_016869940.1 4 1311
GenBank mRNA AI452970.1 - 3 281
AK128289.1 - 2 2529
AL137623.1 - 111
AY358190.1 - 5 603
BC014601.2 - 4 542 4821517
BC041834.1 - 2 2455 5268318
DT220823.1 - 4 561

View Full Product Details