Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_181744.3 | NP_859528.1 | 3 | 419 | |
NR_033806.1 | 2 | 356 | |||
XM_017010410.1 | XP_016865899.1 | 2 | 545 | ||
XM_017010411.1 | XP_016865900.1 | 2 | 527 | ||
XM_017010412.1 | XP_016865901.1 | 2 | 583 | ||
XM_017010413.1 | XP_016865902.1 | 2 | 240 | ||
XM_017010415.1 | XP_016865904.1 | 2 | 356 | ||
XM_017010416.1 | XP_016865905.1 | 3 | 421 | ||
GenBank mRNA | AY288419.1 | - | 3 | 382 | |
AY377391.1 | - | 3 | 396 | ||
BC126194.1 | - | 3 | 419 | 8991910 | |
BC126198.1 | - | 3 | 419 | 8991914 | |
BX647224.1 | - | 2 | 298 | ||
DB056067.1 | - | 2 | 357 |