Gene Transcripts

Gene Symbol
NDUFAF3
Entrez Gene ID
25915
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 3
Gene Aliases
2P1, C3orf60, E3-3
Chromosome Location
Chr. 3: 49020475 - 49023495 on Build GRCh38
UniGene ID
Hs.31387
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199069.1 NP_951032.1 5 1056
NM_199070.1 NP_951033.1 5 619
NM_199073.1 NP_951047.1 5 690
NM_199074.1 NP_951056.1 5 571
GenBank mRNA AL049955.1 - 5 623
BC002873.2 - 5 606 3945015
BC111004.1 - 5 1686 5744973
BQ652614.1 - 5 571

View Full Product Details