Gene Transcripts

Gene Symbol
HEATR4
Entrez Gene ID
399671
Gene Name
HEAT repeat containing 4
Gene Aliases
Chromosome Location
Chr. 14: 73478484 - 73634418 on Build GRCh38
UniGene ID
Hs.660774
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001220484.1 NP_001207413.1 4 1294
NM_203309.2 NP_976054.2 3 1215
XM_006720143.2 XP_006720206.1 3 1252
XM_011536760.2 XP_011535062.1 3 1438
XM_011536761.2 XP_011535063.1 3 1097
XM_017021289.1 XP_016876778.1 3 1776
GenBank mRNA AK093515.1 - 3 1208
AK302564.1 - 4 1268
BC047590.1 - 3 1215 4837558
GU727638.1 - 2 805

View Full Product Details