Gene Transcripts

Gene Symbol
QKI
Entrez Gene ID
9444
Gene Name
QKI, KH domain containing RNA binding
Gene Aliases
Hqk, QK, QK1, QK3, hqkI
Chromosome Location
Chr. 6: 163414486 - 163578596 on Build GRCh38
UniGene ID
Hs.510324
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301085.1 NP_001288014.1 1, 2 697
NM_006775.2 NP_006766.1 1, 2 697
NM_206853.2 NP_996735.1 1, 2 697
NM_206854.2 NP_996736.1 1, 2 697
NM_206855.2 NP_996737.1 1, 2 697
XM_011536258.2 XP_011534560.1 1, 2 204
XM_011536259.2 XP_011534561.1 1, 2 204
XM_011536260.2 XP_011534562.1 1, 2 739
XM_011536261.1 XP_011534563.1 1, 2 204
XM_017011502.1 XP_016866991.1 1, 2 204
XM_017011503.1 XP_016866992.1 1, 2 204
XM_017011504.1 XP_016866993.1 1, 2 204
GenBank mRNA AB067798.1 - 1, 2 698
AB067799.1 - 1, 2 698
AB067800.1 - 1, 2 698
AB067801.1 - 1, 2 698
AF142422.1 - 1, 2 257
AF272349.1 - 1, 2 579
AK027309.1 - 1, 2 104
AY780788.1 - 1, 2 134
BC019917.2 - 1, 2 185 3921310
DQ323998.1 - 1, 2 146

View Full Product Details