Gene Transcripts

Gene Symbol
PLEKHG4B
Entrez Gene ID
153478
Gene Name
pleckstrin homology and RhoGEF domain containing G4B
Gene Aliases
Chromosome Location
Chr. 5: 54065 - 189972 on Build GRCh38
UniGene ID
Hs.535800
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_052909.3 NP_443141.3 16 3295
XM_006714446.3 XP_006714509.1 18 4433
XM_011513968.2 XP_011512270.1 18 4430
XM_011513969.2 XP_011512271.1 18 4310
XM_011513970.1 XP_011512272.1 18 4022
XM_011513971.2 XP_011512273.1 18 4378
XM_017009112.1 XP_016864601.1 18 4434
GenBank mRNA AB067496.2 - 16 3295
AK125163.1 - 5 1124

View Full Product Details