Gene Transcripts

Gene Symbol
KRT77
Entrez Gene ID
374454
Gene Name
keratin 77
Gene Aliases
K1B, KRT1B
Chromosome Location
Chr. 12: 52689612 - 52703524 on Build GRCh38
UniGene ID
Hs.334989
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175078.2 NP_778253.2 9 2230
XM_011538288.2 XP_011536590.1 9 1992
GenBank mRNA AJ564104.1 - 9 2230

View Full Product Details