Gene Transcripts

Gene Symbol
SLC16A6
Entrez Gene ID
9120
Gene Name
solute carrier family 16 member 6
Gene Aliases
MCT6, MCT7
Chromosome Location
Chr. 17: 68267026 - 68291264 on Build GRCh38
UniGene ID
Hs.42645
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001174166.1 NP_001167637.1 3 506
NM_004694.4 NP_004685.2 2 379
XM_005257789.3 XP_005257846.2 2 544
XM_011525461.2 XP_011523763.1 2 223
XM_017025291.1 XP_016880780.1 3 343
XM_017025292.1 XP_016880781.1 2 406
GenBank mRNA AK290417.1 - 2 255
AK313121.1 - 2 245
AX750581.1 - 3 358
AX750582.1 - 1 193
BC064832.1 - 2 381 6498761
U79745.1 - 3 358

View Full Product Details