Gene Transcripts

Gene Symbol
C12orf75
Entrez Gene ID
387882
Gene Name
chromosome 12 open reading frame 75
Gene Aliases
AGD3, OCC-1, OCC1
Chromosome Location
Chr. 12: 105330636 - 105371518 on Build GRCh38
UniGene ID
Hs.368938
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145199.1 NP_001138671.1 6 1150
GenBank mRNA AB735446.1 - 6 1090
AB735447.1 - 5 1114
AJ311378.1 - 6 924
AK056999.1 - 6 1022
BC013920.1 - 6 1152 3838595

View Full Product Details