Gene Transcripts

Gene Symbol
Pold2
Entrez Gene ID
18972
Gene Name
polymerase (DNA directed), delta 2, regulatory subunit
Gene Aliases
50kDa, p50, po1D2
Chromosome Location
Chr. 11: 5872180 - 5878256 on Build GRCm38
UniGene ID
Mm.35788
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008894.2 NP_032920.2 8 1035
GenBank mRNA AK013901.1 - 8 1001
BC003342.1 - 8 1001 3257610
CB205181.1 - 5 571
Z72486.1 - 8 996

View Full Product Details