Gene Transcripts

Gene Symbol
Sms
Entrez Gene ID
20603
Gene Name
spermine synthase
Gene Aliases
AI427066, Gy, SPMSY, SpmST, gyro
Chromosome Location
Chr. X: 157443852 - 157492046 on Build GRCm38
UniGene ID
Mm.18652
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009214.3 NP_033240.3 4, 5 428
GenBank mRNA AF031486.1 - 4, 5 411
AK002687.1 - 4, 5 426
AK017775.1 - 4, 5 399
AK049780.1 - 4, 5 427
AK049787.1 - 4, 5 417
AK136798.1 - 4, 5 429
AK149929.1 - 4, 5 426
AK160659.1 - 4, 5 429
AK166665.1 - 4, 5 454
BC046623.1 - 4, 5 410 6315420
BC058688.1 - 4, 5 669 6413869
Y09419.1 - 4, 5 401

View Full Product Details