Gene Transcripts

Gene Symbol
Prl7d1
Entrez Gene ID
18814
Gene Name
prolactin family 7, subfamily d, member 1
Gene Aliases
PLF-RP, PRP, Plfr
Chromosome Location
Chr. 13: 27708998 - 27716748 on Build GRCm38
UniGene ID
Mm.3258
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011120.2 NP_035250.1 2 253
GenBank mRNA AK005421.1 - 2 250
AK005557.1 - 2 253
AK019461.1 - 2 253
BC099470.1 - 2 242 30787403
X02594.1 - 2 253

View Full Product Details