Gene Transcripts

Gene Symbol
Apcs
Entrez Gene ID
20219
Gene Name
serum amyloid P-component
Gene Aliases
Sap
Chromosome Location
Chr. 1: 172893961 - 172895054 on Build GRCm38
UniGene ID
Mm.330510
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011318.2 NP_035448.2 2 351
GenBank mRNA AK144248.1 - 2 238
AK165699.1 - 2 238
BC061125.1 - 2 248 30308397
M23552.1 - 2 327
X14079.1 - 2 327
Y00426.1 - 2 340

View Full Product Details