Gene Transcripts

Gene Symbol
Slc12a7
Entrez Gene ID
20499
Gene Name
solute carrier family 12, member 7
Gene Aliases
AA408796, D13Ertd261e, Kcc4
Chromosome Location
Chr. 13: 73733053 - 73816755 on Build GRCm38
UniGene ID
Mm.275800
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011390.2 NP_035520.1 17 2279
GenBank mRNA AF087436.1 - 17 2281
AK131129.1 - 17 2233
AK143535.1 - 17 2273
AK149750.1 - 17 2274
AK166215.1 - 17 2273
AK169950.1 - 17 2269
AK171498.1 - 17 2316
BC059242.1 - 17 2271
BC060133.1 - 17 2271
BC079677.1 - 14 4029
BC141107.1 - 17 2239 9055719

View Full Product Details