Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011481.3 | NP_035611.3 | 5 | 1319 | |
GenBank mRNA | AK049582.1 | - | 5 | 580 | |
BC120633.1 | - | 5 | 1298 | 40129556 | |
BC125325.1 | - | 5 | 1295 | 40129840 | |
D26186.1 | - | 5 | 1165 | ||
D49427.1 | - | 5 | 1333 |